missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ beta-Defensin 1 Human ELISA Development Kit (ABTS), PeproTech®, PeproTech®
ELISA
Brand: Invitrogen™ 900-K202
Additional Details : Weight : 0.03500kg
Description
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 1, an antimicrobial peptide implicated in the resistance of epithelial surfaces to microbial colonization. This gene maps in close proximity to defensin family member, defensin, alpha 1 and has been implicated in the pathogenesis of cystic fibrosis.Specifications
P60022 | |
Lot-specific | |
ELISA | |
1672 | |
This Human BD-1 ELISA Kit includes: Capture Antibody, Detection Antibody, Protein Standard, and HRP-Conjugate. | |
10 x 96 Tests | |
Refer to product documentation for component specific storage temperature. | |
1 hr 45 min |
Lot-specific | |
Biotin | |
Colorimetric Microplate Reader | |
DEFB1 | |
HRP | |
RUO | |
Human | |
24 hr 30 min |