Learn More
Cytiva Benchkote sheet for ÄKTA systems
Benchkote sheets for ÄKTA systems will protect the top buffer trays from buffer spillages and salt deposits.
Brand: Cytiva 2300-10094
42.13 EUR valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 0.55000kg
Description
Why Benchkote sheet for lab ÄKTA systems?
- Cover the top buffer tray with Benchkote sheet and protect from buffer spillages and salt deposits.
- Simplify daily and weekly preventive maintenance of the top buffer tray of your ÄKTA systems
- Smooth, absorbent Whatman™ paper quickly absorbs spills.
- Laminated polyethylene layer prevents flow-through to the working surface.
- Offered in three convenient pack sizes: 10/pk, 25/pk and 50/pk sheets.
- Convenient and easy to use.
- Benchkote sheets are manufactured of absorbent, impermeable material designed to protect lab surfaces against hazardous spills.
- The high-quality, smooth, and absorbent Whatman™ paper, quickly absorbs liquid spills, and laminated polyethylene layer prevents flow-through to the working surface.
Specifications
Absorbent Paper, Laminated Polyethylene | |
For use with ÄKTA go | |
25/Pk. |