Learn More
Bel-Art™ Write-On™ Paper Label Tapes
Product Code. p-4520245
Change view
Click to view available options
Color:
Assorted
Blue
Green
Orange
Red
White
Yellow
Dimensions (L x W):
13.7 m x 19 mm
13.7 m x 25.4 mm
36.4 m x 1.9 cm
36.4 m x 12.7 mm
36.4 m x 19 mm
36.4 m x 25.4 mm
36.4 m x 25.4mm
54.6 m x 12.7 mm
54.6 m x 19 mm
Diameter (English):
1 in.
3 in.
Unit Size:
Pack of 3
Pack of 4
Pack of 6
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Dimensions (L x W) Diameter (English) unitSize
13455236 Orange 36.4 m x 12.7 mm 3 in. Pack of 6
13465236 Blue 36.4 m x 12.7 mm 3 in. Pack of 6
13405246 Blue 36.4 m x 19 mm 3 in. Pack of 4
13435236 Red 36.4 m x 12.7 mm 3 in. Pack of 6
13495236 Yellow 36.4 m x 19 mm 3 in. Pack of 4
13455226 White 13.7 m x 19 mm 1 in. Pack of 4
13475246 Blue 36.4 m x 25.4 mm 3 in. Pack of 3
13415246 Green 36.4 m x 19 mm 3 in. Pack of 4
13474166 White 36.4 m x 12.7 mm 3 in. Pack of 6
10045180 White 54.6 m x 19 mm 3 in. Pack of 4
13425246 Orange 36.4 m x 19 mm 3 in. Pack of 4
13475236 White 36.4 m x 19 mm 3 in. Pack of 4
13405236 White 13.7 m x 25.4 mm 1 in. Pack of 3
13435246 White 36.4 m x 25.4mm 3 in. Pack of 3
13445246 Red 36.4 m x 25.4 mm 3 in. Pack of 3
13465246 Green 36.4 m x 25.4 mm 3 in. Pack of 3
13485236 Red 36.4 m x 19 mm 3 in. Pack of 4
15196324 Assorted 36.4 m x 1.9 cm 3 in. Pack of 6
13485246 Orange 36.4 m x 25.4 mm 3 in. Pack of 3
13495246 White 54.6 m x 12.7 mm 3 in. Pack of 6
13434720 Yellow 36.4 m x 12.7 mm 3 in. Pack of 6
13455246 Yellow 36.4 m x 25.4 mm 3 in. Pack of 3
13445236 Green 36.4 m x 12.7 mm 3 in. Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Product Code. 13455236 Supplier Bel-Art™ Supplier No. F134880050
Only null left
Add to basket
Edge
Add to basket
Use pencil, ball-point pen, or solvent ink marker to create labels that remain legible under the most demanding laboratory conditions. Paper label tapes resist moisture and most solvents and are great for use in low temperature applications.
  • Adheres to any clean surface, including PTFE fluoropolymer resin coated materials
  • Peels off without leaving a sticky residue
  • Pencil, ball-point pen, or solvent ink marker remain legible under the most demanding laboratory conditions
  • Colored paper with rubber resin adhesive resists moisture and most solvents
  • Withstands a maximum temperature of 157°C (315°F) for 30 minutes or 80°C (176°F) for 24 hours; can be used down to -80°C (-112°F)
  • Withstands a steam autoclaving cycle at 121°C (250°F) and gamma irradiation between 13.0 and 45.0kGy
Spezifikation
Autoclavable Autoclavable
Adhesive Type Rubber, Resin
Coating None
Color Orange
Display Blank
Finish Matte
Format Roll
Material Paper
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Ultra Reflective Letter Label
Sterility Non-sterile
For Use With (Application) For use in low temperature applications
Quantity 6/Pk.
Dimensions (L x W) 36.4 m x 12.7 mm
Length (English) 40 yd.
Length (Metric) 36.5 m
Width (English) 0.5 in.
Width (Metric) 12.7 mm
Diameter (English) 3 in.
Diameter (Metric) 7.62 cm
Mehr anzeigen Weniger anzeigen