Learn More
BD Rat C-reactive protein (CRP) ELISA Kit
For detection and quantitation of rat CRP in rat serum
Brand: BD 557825
Additional Details : Weight : 0.10000kg Transport : UN number : 1805 Chem class : 8 Pack group : III
Includes: Capture antibody coated microplate, detection antibody/enzyme conjugate (100x), standard (10x), wash buffer, TMB substrate, stop solution
Description
The C-reactive protein is an acute-phase protein produced by liver during conditions of inflammation, bacterial infection, or tissue trauma
- Reactivity: Rat
- Useful for determination of inflammatory conditions
- Utilizes an antibody specific for rat CRP coated on a 96-well plate
Enzyme-linked immunosorbent assay
Specifications
CRP | |
Rat C-Reactive protein ELISA microplate (96-well coated with Anti-CRP), Rat C-Reactive protein standard (10X), HRP Conjugated Rabbit Anti-rat CRP (100X), PBS-T Wash Buffer powder, TMB Substrate, stop solution | |
Rat Serum | |
Detection and quantitation of Rat CRP in Rat Serum, ELISA |
Rat | |
1 Plate | |
2°C to 8°C | |
Rat C-reactive Protein ELISA kit |
Research Use Only