Learn More
BD OptEIA™ Monkey IFN-γ ELISA Set
For natural or recombinant Interferon-γ (IFN-γ) in non-human primate (rhesus, cynomolgus, and baboon) serum, plasma and cell culture supernatants.
Brand: BD 551492
Additional Details : Weight : 0.16000kg
Description
Standard in this set is calibrated against purified Baculovirus-expressed recombinant human IFN-γ. One monkey IFN-γ unit (U) is defined as one picogram of recombinant human IFN-γ.
- Kit contents: capture antibody (Anti-Monkey IFN-γ monoclonal antibody), detection antibody (Biotinylated Anti-Monkey IFN-γ monoclonal antibody), enzyme reagent (Streptavidin-horseradish peroxidase conjugate SAv-HRP), standards (Recombinant IFN-γ, lyophilized), instruction/analysis certificate
- Size: 20 tests of 96-well format
ELISA
Specifications
Monkey IFN-g ELISA set | |
To develop ELISA for natural or recombinant IFN-g in non-Human Primate (rhesus,cynomolgus and baboon) Serum, Plasma and cell culture supernatants | |
20 Plates |
20 96-well Plates | |
anti-Monkey IFN-g monoclonal (capture) antibody, Biotinylated anti-Monkey IFN-g monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant IFN-g (standard), Streptavidin-HRP | |
2°C to 8°C |