missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BV480 Rat Anti-Mouse CD117
Widespread expression of CD117 in hematopoietic precursors is consistent with participation of c-Kit and its ligand in regulation of several hematopoietic lineages. BD Horizon™ BV480 Rat Anti-Mouse CD117 is also involved in the regulation of some events during the development of T lymphocytes. CD117 is also expressed by mast cells and by dendritic cells found in the periarteriolar lymphocytoc sheaths (T-cell areas) of splenic white pulp.
297.00€ - 765.00€
Specifications
Product Type | BV480 Rat Anti-Mouse CD117 |
---|---|
For Use With (Application) | Flow Cytometry |
Concentration | 0.2 mg/mL |
Cross Reactivity | QC Testing: Mouse |
Form | BV480 |
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Rat Anti-Mouse CD117 | |
0.2 mg/mL | |
BV480 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
Flow Cytometry | |
QC Testing: Mouse | |
Store undiluted at 4°C |