Learn More
BD BV480 Mouse Anti-Human CD8
CD8 is expressed by majority of thymocytes, by subpopulations of αβ T cells and γδ T cells and by some NK cells. BD Horizon™ BV480 Mouse Anti-Human CD8 plays important roles in T cell activation and selection.
Brand: BD 566121
Additional Details : Weight : 0.01000kg
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Mouse Anti-Human CD8 | |
Flow Cytometry | |
BV480 | |
Store undiluted at 4°C |
100 Tests | |
QC Testing: Human; Tested in Development: Rhesus, Cynomolgus, Baboon | |
5 μL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |