missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BV480 Mouse Anti-Human CD31
CD31 has wide tissue distribution and is expressed on platelets, monocytes, granulocytes, NK cells, T cell subsets, and in high amounts on endothelial cells. BD Horizon™ BV480 Mouse Anti-Human CD31 functions as a vascular endothelial cell adhesion molecule and is involved in the transendothelial migration of leucocytes in inflammatory responses.
316.00€ - 1120.00€
Specifications
Product Type | BV480 Mouse Anti-Human CD31 |
---|---|
For Use With (Application) | Flow Cytometry |
Cross Reactivity | QC Testing: Human |
Form | BV480 |
Sample Volume | 5 μL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15874829
|
BD
566195 |
25 Tests |
316.00€
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15894779
|
BD
566144 |
100 Tests |
1120.00€
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Mouse Anti-Human CD31 | |
QC Testing: Human | |
5 μL | |
5175 |
Flow Cytometry | |
BV480 | |
Store undiluted at 4°C | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |