Learn More
BD Cytometric Bead Array (CBA) Human IL-2 Enhanced Sensitivity Flex Set (Bead A4)
Measures human Interleukin-2 (IL-2) in serum, plasma and cell culture supernatant samples
Brand: BD 561517
Additional Details : Weight : 0.04000kg
Alert:
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.
Includes: Kit contents:
Enhanced Sensitivity Capture Bead A4: stored in aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
Detection Reagent (Part A): stored in aqueous buffered solution containing BSA and ProClin™ 150
Standard: lyophilized in an aqueous buffered solution containing BSA and ProClin 150
Description
This kit is used in conjunction with BD™ Cytometric Bead Array (CBA) Human Enhanced Sensitivity Master Buffer Kit, flow cytometer and FCAP Array™ Software and can measue soluble analytes present in very low concentrations.
- Bead-based immunoassay
- Reactivity: human
- Standard assay range: 274 to 200000fg/mL
Flow Cytometry/Immunoassay
Specifications
IL-2 | |
1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized) | |
Serum, Plasma, Cell Culture Supernatant | |
Measures Human interleukin-2, flow cytometry and immunoassay |
Human | |
100 Tests | |
4°C | |
Human IL-2 enhanced sensitivity flex set (bead A4) |
Safety and Handling
- BD Cytometric Bead Array (CBA) Human IL-2 Enhanced Sensitivity Flex Set
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 5-chloro-2-methyl-4-isothiazolin-3-one