Learn More
BD Cell Signaling Master Buffer Kit
Employ particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations
Brand: BD 560005
Additional Details : Weight : 0.60000kg
Includes:
- Assay diluent
- Capture bead diluent
- Detection reagent diluent
- Denaturation buffer
- Wash buffer
- Instrument setup reagents
Description
The buffers and instrument setup reagents provided in this kit have been optimized to measure total or phosphorylated proteins from denatured cell lysate sample.
- Contains all of the supporting reagents necessary to perform an assay using a BD CBA Cell Signaling Flex Set
- Provides sufficient reagents for the quantitative analysis of 100 samples and 10 instrument setup procedures
- Working assay range for most analytes in this system is 3.9 to 1,000 units/mL
For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
1 bottle 30mL assay diluent, 1 bottle 5mL capture bead diluent, 1 bottle 5mL detection reagent diluent, 1 bottle 30mL denaturation buffer 5X, 1 bottle 130mL wash buffer, 1 vial 0.25mL instrument setup bead A1, 1 vial 0.25mL instrument setup bead A9 | |
100 | |
RUO |
2°C to 8°C | |
Cytometric Bead Array (CBA) Assay |