Learn More
Thermo Scientific™ Barnstead™ Deionization System Disposable Filters
Accessory/replacement consumable for Nanopure™ (Analytical, Analytical UV, Biological UF, Life Science UV/UF), Easypure™ II (UV, UF, UV/UF), Easypure II RF (UV), Easypure RoDi, E-Pure™ (3,4 holder), TII
Brand:  Thermo Scientific™ FL703X2
Additional Details : Weight : 0.02160kg
 View more versions of this product
Product Code. 10757074
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
Accessory/replacement consumable for Nanopure™ (Analytical, Analytical UV, Biological UF, Life Science UV/UF), Easypure™ II (UV, UF, UV/UF), Easypure II RF (UV), Easypure RoDi, E-Pure™ (3,4 holder), TII
Specifications
Specifications
Autoclavable
0.2 μm
Filling bell
1
Reverse osmosis and TII water purification system
Deionization System Disposable Filter