Learn More
Thermo Scientific™ Barnstead™ 1/2 Size B-Pure™ Filters
Pre-treat your feed water with Thermo Scientific™ Barnstead™ 1/2 Size B-Pure™ Filters, available in varying pore sizes.
84.80€ - 393.00€
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12903311
|
Thermo Scientific™
FL583X1 |
5µm Pre-filter for 1/2 Size B-Pure Filter Holder |
84.80€
Each |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||||
15395985
|
Thermo Scientific™
FL583X2 |
10µm Pre-filter for 1/2 Size B-Pure Filter Holder |
86.80€
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
12913311
|
Thermo Scientific™
FL583X3 |
15µm Pre-filter for 1/2 Size B-Pure Filter Holder |
84.80€
Each |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
15322188
|
Thermo Scientific™
FL583X4 |
1µm Pre-filter for 1/2 Size B-Pure Filter Holder |
86.80€
1 unit |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
11830880
|
Thermo Scientific™
FL583X5 |
0.45µm Final Filter for 1/2 Size B-Pure Filter Holder |
380.00€
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
12923311
|
Thermo Scientific™
FL583X6 |
0.2µm Final Filter for 1/2 Size B-Pure Filter Holder |
393.00€
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
Description
Pre-treat your feed water with Thermo Scientific™ Barnstead™ 1/2 Size B-Pure™ Filters. In addition to pre-treating feed water, the filters can act as a final filter to ensure consistently pure water. Compatible with B-Pure 1/2 size filter holders, these filters are available in varying pore sizes to meet all of your pre-filtration needs.
- Large surface area for long life
- Flow rates of up to 2L/min.