Learn More
Masterflex™ Barbed Reducer
Combine these simple, economical fittings with flexible tubing for use in low-pressure applications (generally less than 30 psi). Cole-Parmer™ Barbed Reducers offer very good chemical resistance. No tools needed for assembly.
Brand: Masterflex™ 30622-27
Additional Details : Weight : 0.01128kg
Description
- Temperature range: –53 to 65°C
- Sterilize by ethylene oxide only
Specifications
Fits 1/4 in. x 1/8 in. I.D tubing | |
10/Pk. | |
65°C |
-53°C | |
Straight |