Learn More
J.T. Baker Baker-flex™, Silica Gel IB2, J.T. Baker™
Flexible TLC Sheet
Brand:  J.T. Baker 4448-04
807.50 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Additional Details : CAS : 7631-86-9 Weight : 0.01000kg
 View more versions of this product
Product Code. 13295423
  • 850.00€ / Pack of 25
Estimated Shipment: 04-09-2024
Description
Flexible TLC Sheet
Specifications
Specifications
White
25 Sheets (20 x 20 cm) 10/case
Silica Gel IB2