Learn More
Azlon™ Square Shaped HDPE Bucket
Square shaped buckets suitable for food contact applications in accordance with EU regulations
Brand:  Azlon BWX104
39.38 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.62000kg
 View more versions of this product
Product Code. 12015562
  • 41.45€ / Each
Estimated Shipment: 16-07-2024
Description
  • Durable square shape allows economical use of space
  • Handle grip for easy lifting and carrying
  • Molded graduations (14L x 0.5 divisions)
Specifications
Specifications
Spouted Bucket
0.5 L
Black
283 mm
10.5 L
HDPE, Handle: Polyethylene
286 mm