Learn More
Axygen™ 1.5 mL Conical Screw Cap Tubes
Product Code. 11370375 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Closure Color:
Assorted
Clear
Red
Unit Size:
Pack of 4000
Pack of 500
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Closure Color Unit Size
11370375 Non-sterile Clear Pack of 500
11340375 Sterile Assorted Pack of 500
12176830 Non-sterile Clear Pack of 4000
11380375 Sterile Clear Pack of 500
11330385 Non-sterile Red Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11370375 Supplier Axygen™ Supplier No. SCT150C
  • ONLINE EXCLUSIVE PRICE
    163.00€
    108.93€ / Pack of 500
    Save 54.07€
    33% Off
In Stock
Add to basket
Edge
Add to basket
With chemical-resistant O-ring
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
  • Available as presterilized and nonsterile
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order
  • Caps available in range of colors for easy sample identification
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Clear
Closure Material Polypropylene
Capacity (Metric) 1.5 mL
Color Clear
Material Polypropylene
Format Non-looped
Includes O-ring
Sterility Non-sterile
Graduated No
Closure Type Screw Cap
Bottom Shape Conical
Max. RCF 20,000 x g
Product Type Microcentrifuge Tube