Learn More
Trajan Scientific and Medical™ Autosampler Syringes with Cone Tipped Needle for Thermo Scientific GCs
Gives better performance and superior quality. Trajan Scientific and Medical Autosampler Syringes with Cone Tipped Needle for Thermo Scientific GCs has the knowledge and expertise to manufacture an SGE syringe.
109.00€ - 142.00€
Specifications
Needle Gauge | 26 G |
---|---|
Diameter (Metric) Outer Needle | 0.47 mm |
Needle Point Style | Cone |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15218443
|
Trajan Scientific and Medical™
000790 |
2 μL |
142.00€
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
15218473
|
Trajan Scientific and Medical™
0029931 |
10 μL |
109.00€
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Thermo Scientific instruments
Specifications
26 G | |
0.47 mm | |
Cone |