missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint-Gobain AstiPure™ TP1 Series PFA Union Tee Fittings
60.45€ - 65.65€
Specifications
Product Type | Union Tee Fittings |
---|---|
Material | PFA |
Packaging Quantity | 1 Each |
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
11984099
|
Saint-Gobain
TP10608 |
5 mm |
65.65€
Each |
In Stock
Log in to see stock. |
|||||
11999934
|
Saint-Gobain
TP10406 |
4 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
Union Tee Fittings | |
PFA | |
1 Each |