Learn More
Saint-Gobain AstiPure™ RC1 Series PFA Union Panel Mount
Brand:  Saint-Gobain RC10406
18.97 EUR valid until 2024-05-31
Use promo code "21638" to get your promotional price.
Additional Details : Weight : 0.01000kg
Product Code. 11944139
  • 48.85€ / Each
In Stock
to see stock.
Add to basket
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Specifications
Union Panel Mount
58 mm
1 Each
PFA
For 4 x 6mm dia. tubing
12.5 x 5mm