missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Nitrate, ACS Reagent, Honeywell Fluka™

Catalog Number p-7071892
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
6484-52-2
H4N2O3
80.04
MFCD00011425
DVARTQFDIMZBAA-UHFFFAOYSA-O
ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
22985
CHEBI:63038
ammonium nitrate
[NH4+].[O-][N+]([O-])=O
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
6484-52-2
H4N2O3
80.04
MFCD00011425
DVARTQFDIMZBAA-UHFFFAOYSA-O
ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
22985
CHEBI:63038
ammonium nitrate
[NH4+].[O-][N+]([O-])=O
Quantity 500 g
Linear Formula NH4NO3
UN Number 1942
Formula Weight 80.04g/mol
Percent Purity ≥98%
Grade ACS Reagent
Residue after Ignition ≤0.01%
Chemical Name or Material Ammonium Nitrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'