Learn More
Cytiva Amersham™ Hyperfilm™ MP
Use in automatic processors or manually in tanks and trays using most common X-ray film developers and fixers. Cytiva Life Sciences™ Amersham™ Hyperfilm™ MP is an excellent multipurpose film constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti scratch layer.
261.00€ - 660.00€
Specifications
For Use With (Application) | 32P, 33P, 125I Southern, Northern, and Western blots. 32S, 33P, 35P sequencing. Fluorography of 3H, 14C, and 35S |
---|
Product Code | Brand | Dimensions (LxW) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | Quantity | Price | Quantity & Availability | ||||
10627265
|
Cytiva
28-9068-46 |
8 x 10 in. | 100 Sheets/Box, Single film bag |
660.00€
100 sheets |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
10742837
|
Cytiva
28-9068-45 |
8 x 10 in. | 50 Sheets/Box, Single film bag |
461.30€
50 sheets |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
10034944
|
Cytiva
28-9068-42 |
5 x 7 in. | 50 Sheets/Box, Single film bag |
261.00€
50 sheets |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
10094984
|
Cytiva
28-9068-44 |
18 x 24 cm | 100 Sheets/Box, Single film bag |
496.00€
100 sheets |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
10218674
|
Cytiva
28-9068-43 |
18 x 24 cm | 50 Sheets/Box, Single film bag |
307.00€
50 sheets |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
10084764
|
Cytiva
28-9068-50 |
18 x 24 cm | 50 Sheets/Box, Individual film envelope |
315.00€
50 sheets |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
Description
- High sensitivity for low concentration targets
- Optimized for direct and indirect autoradiography with 3H, 14C, 32P, 33P, 35S, 125I
- Clear background for excellent contrast and improved band visibility.
- Publication-quality images
- High resolution at an economical price
- Easy to use
Specifications
32P, 33P, 125I Southern, Northern, and Western blots. 32S, 33P, 35P sequencing. Fluorography of 3H, 14C, and 35S |