Learn More
Amberlite™ IRN-150, ion exchange resin, Thermo Scientific Chemicals
Styrene-DVB; Mix of strongly acidic and basic gel type resin
Brand: Thermo Scientific Alfa Aesar 40992-A9
Additional Details : Weight : 10.00000kg
Description
Amberlite IRN-150 is used in water treatment and purification in primary side chemical volume control systems (CVS) in PWR nuclear power generating facilities. It is also used in reactor water clean-up and radio active waste systems. It is recommended in any non-regenerable mixed bed application and can also be used for a variety of electroplating industry waste regeneration applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Beads | |
Odorless | |
MFCD00145822 | |
Amber | |
Insoluble in water,acids and bases. |
Amberlite IRN-150 | |
Styrene-DVB | |
1.1-1.3g/cm3 | |
10 kg |
RUO – Research Use Only