Learn More
Amberlite™ IRN-150, ion exchange resin, Thermo Scientific Chemicals
Styrene-DVB; Mix of strongly acidic and basic gel type resin
Brand:  Thermo Scientific Alfa Aesar 40992-A9
Additional Details : Weight : 10.00000kg
 View more versions of this product
Product Code. 16515500
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
Amberlite IRN-150 is used in water treatment and purification in primary side chemical volume control systems (CVS) in PWR nuclear power generating facilities. It is also used in reactor water clean-up and radio active waste systems. It is recommended in any non-regenerable mixed bed application and can also be used for a variety of electroplating industry waste regeneration applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Beads
Odorless
MFCD00145822
Amber
Insoluble in water,acids and bases.
Amberlite IRN-150
Styrene-DVB
1.1-1.3g/cm3
10 kg
Product Suggestions
Product Suggestions
Videos
SDS Documents