Learn More
Bochem™ Aluminum Universal Clamp
Product Code. 10622611
Change view
Click to view available options
Diameter (Metric):
0 to 80 mm
Length (Metric) Arm:
180 mm
Material:
Aluminum
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) Arm Material Unit Size
10622611 0 to 80 mm 180 mm Aluminum Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10622611 Supplier Bochem™ Supplier No. 5444
  • ONLINE EXCLUSIVE PRICE
    35.15€
    20.83€ / Pack of 1
    Save 14.32€
    40% Off
In Stock
Add to basket
Edge
Add to basket
Manufactured from aluminum. BOCHEM™ Aluminum Universal Clamp is DIN 12895 certified.
  • Finger with cork
  • Aluminum
  • DIN 12895
Specifications
Quantity 1 Ea.
Material Aluminum
Diameter (Metric) 0 to 80 mm
Length (Metric) Arm 180 mm