Learn More
R&L Enterprises™ Retort Stand Clamps
Product Code. 11785376 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Size (Metric) Grip:
85 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) Grip Unit Size
11785376 85 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11785376
Estimated Shipment: 19-01-2027
Add to basket
Add to basket
Enables articles from 2 to 90mm in diameter to be clamped securely. R&L Enterprises™ Retort Stand Clamps feature aluminum alloy jaws with other parts of plated brass.
  • Design ensures a firm grip at all angles of opening
  • Cork liners for the jaws are well secured
Specifications
Type Retort Clamp
Material Aluminum
Includes Cork-lined Jaws
For Use With (Equipment) For use with 85mm diameter
Size (Metric) Grip 85 mm
Jaw Type Four Interlocking