Learn More
Brady™ Aluminum Laser Radiation Warning Signs
6.10€ - 25.30€
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
11780284
|
Brady™
241595 |
100 mm |
6.10€
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
11790284
|
Brady™
241597 |
315 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
11826371
|
Brady™
241596 |
200 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
Triangular |