Learn More
Bochem™ Aluminum Laboratory Pot With Lid
Brand:  Bochem™ 8567
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10044920
  • 39.10€ / Each
Estimated Shipment: 17-01-2023
Specifications
3 L
Aluminum
Lid
Laboratory Pot
120 mm
200 mm