Learn More
Ansell AlphaTec™ 87-108 Heavy Duty Chemical Protection Glove
Product Code. 17225106 Shop All Ansell Products
Change view
Click to view available options
Size:
7.5
8.5
9.5
10.5
Unit Size:
1 pair
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17225106 8.5 1 pair
17215106 7.5 1 pair
17235106 9.5 1 pair
17245106 10.5 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17225106 Supplier Ansell Supplier No. 87108085
  • ONLINE EXCLUSIVE PRICE
    52.20€
    33.53€ / 1 pairs
    Save 18.67€
    35% Off
In Stock
Add to basket
Edge
Add to basket
AlphaTec™ 87-108 Heavy Duty Chemical Protection Gloves feature good mechanical and chemical protection in heavy duty environments
  • High resistance to water-based chemicals
  • Good flexibility and elasticity preventing tears
  • Chlorinated to harden and cleanse the surface of the glove
  • AQL 0.65
TRUSTED_SUSTAINABILITY
Specifications
Applications Industrial
Color Black
Latex Free No
Material Rubber
Palm Thickness (English) 0.0591 in.
Palm Thickness (Metric) 1.5 mm
Product Type Chemical Protection Gloves
Size 8.5
Thickness (English) 59 mil
Thickness (Metric) 1.5 mm