Description
- Specialized protection: AlphaTec™ 4000 CFR Stitched and Taped - Model 111 chemical protective suit features multi-hazard solutions that offer a proven 8 hour barrier against chemicals such as non-flammable, flammable and aggressive solvents
- Improved durability: Complying with EN ISO 14116 and ASTM D6413 Flame Resistance*, this full protective suit shields the wearer from chemical sprays without compromising safety by limiting flame spread in the event of a flash fire
- Elevated comfort: The optimized body fit design of this protective suit offers improved wearer comfort and safety
- This full-chemical protection suit defends against infective agents, tested according to ASTM F1671 and hydrocarbons such as hexane, HF, oleum and many more
- *AlphaTec™ 4000 CFR should never be worn in isolation for flame retardant protection.
- Always wear over the top of garments which achieve EN ISO 14116 Index 2 or above
- Polyolefin-based core barrier laminate, with FR non-woven inner and FR outer barrier film
Specifications
Specifications
| Product Type | Chemical-resistant Coveralls |
| Size | Large |
| Gender | Unisex |
| Material | Polyolefin |
| Color | Orange |
| Disposable | Single-use |
| Waist Style | Elastic at Back |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Hood Type | Attached |
| Show More |