Learn More
Texwipe™ AlphaSorb™ 10 Sealed-Border Polyester Wipers
BIOPHARMA
Product Code. 15684977 Shop All Texwipe Products
Change view
Click to view available options
Dimensions (L x W):
12 x 12 in.
9 x 9 in. (22.86 x 22.86 cm)
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 500
Pack of 600
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Sterility Unit Size
15684977 12 x 12 in. Sterile Pack of 500
15702396 9 x 9 in. (22.86 x 22.86 cm) Non-sterile Pack of 100
18603943 12 x 12 in. Non-sterile Pack of 600
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15684977 Supplier Texwipe™ Supplier No. TX3215S
  • ONLINE EXCLUSIVE PRICE
    1114.95€
    951.70€ / Pack of 500
    Save 163.25€
    14% Off
In Stock
Add to basket
Edge
Add to basket
For ISO Class 3 and 4 applications
  • Double-knit, no-run interlock construction of 100% continuous-filament polyester
  • Two-ply, sealed-border wiper with highest sorbency
  • Ideal wiper for use in multiple environments
  • Excellent for picking up large amounts of liquids and for applying chemicals
  • Abrasion-resistant wiper with soft, flexible hand
  • Lot-to-lot traceability
Order Info
Available dry or sterile. Basis weight: 185g/m2.
Specifications
Product Type Cleanroom Wipe
Length (English) 12 in.
Length (Metric) 30.5 cm
Width (English) 12 in.
Width (Metric) 30.5 cm
Material Polyester
Basis Weight 185
Certifications/Compliance ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
Edges Sealed
Dimensions (L x W) 12 x 12 in.
For Use With (Application) Ideal wipes for use in multiple environments