Learn More
BTX™ AgilePulse PLUS Safety Stand
Adjustable Gap Safety Stand, specifically for AgilePulse PLUS Electroporators and BTX Flatpack chambers.
Brand: BTX™ 470610N
Additional Details : Weight : 0.90000kg
Description
- Connect to Agilepulse PLUS for safe delivery of high voltage electrical pulses to BTX Flatpack chambers
- For optimum transfections up to volumes of 10 mL (Part Number 47-0206)
Specifications
Large Volume Transfection | |
110/240 V | |
Touchscreen | |
Electrode Connector Cable | |
Square Wave |
AgilePulse PLUS Electroporator | |
AgilePulse PLUS Safety Stand for Flatpack | |
Non-sterile | |
Digital User |