Learn More
FTM Technologies™ Active Carbon Filters
Used to eliminate noxious or unpleasant smells emanating from liquids, vapours or smoke. FTM Technologies™ Active Carbon Filters are for use with fume hoods.
83.65€ - 173.00€
Specifications
Material | Activated Carbon |
---|---|
For Use With (Application) | For Organic Vapors, Acids and Odors |
No. per Pack | 1Each |
Type | Filter |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11738044
|
FTM Technologies™
FCP1 |
Fume Hoods (Except for HI8F/HI8V) |
173.00€
Each |
In Stock
Log in to see stock. |
|||||
11738024
|
FTM Technologies™
FC2 |
HI8F/HI8V Fume Hoods |
83.65€
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Description
- Superior protection in the laboratory
- Compatible with a wide range of fume hoods
Specifications
Activated Carbon | |
For Organic Vapors, Acids and Odors | |
1Each | |
Filter |