missing translation for 'onlineSavingsMsg'
Learn More

Rogo Sampaic™ Acrylic Rack

Brand:  Rogo Sampaic™ BJB032

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11758512

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

Test Tube Rack
Tubes (30 mm)
6
1 Each
70 mm
30 mm
2 x 3
Acrylic
32mm Diameter Tube
140 mm
95 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers