missing translation for 'onlineSavingsMsg'
Learn More

Rogo Sampaic™ Acrylic Rack

Brand:  Rogo Sampaic™ BJB027

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11732724

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »


Description

Description

Specifications

Specifications

Test Tube Rack
Tubes (14 mm)
12
1 Each
50 mm
14 mm
2 x 6
Acrylic
14mm Diameter Tube
105 mm
55 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers