Learn More
Cytiva (formerly Pall Lab) AcroPrep™ 384-well Filters Plates, 100 μL
AcroPrep™ 384-well Filter Plates are ideal for use in high volume, high throughput sample prep applications.
Brand: Cytiva (formerly Pall Lab) 5085
Additional Details : Weight : 0.77900kg
Description
- Plates are constructed from chemically-resistant, biologically-inert polypropylene with a clear polystyrene lid
- This product is for research applications only
Specifications
Natural | |
Supor (PES) | |
Non-sterile | |
Short Tips |
Polypropylene | |
1.2 μm | |
10/Pk. | |
80 μL |