Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with GF/Nylon Membrane
Versatile Filter for both aqueous and solvent-based sample filtration.
Brand:  Cytiva (formerly Pall Lab) 4549
169.11 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Additional Details : Weight : 1.18750kg
Product Code. 17154351
  • 185.00€ / Pack of 50
In Stock
Description
  • Excellent chemical compatibility with esters, bases, and alcohols
  • Saves time because no prewetting is required
Specifications
Specifications
Nylon
0.45 μm
Non-sterile
25 mm