missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Accucore™ 150-C4 HPLC Columns

Use this specially designed solid core column with a C4 functionality for biomolecules that bind too tightly to C8 or C18 columns.

Brand:  Thermo Scientific™ 16526-032130

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12685695

  • 589.07€ / Each
In Stock
to see stock.
Add to basket

Description

Description

Offer lower hydrophobic retention, ideal for optimal retention of proteins and larger peptides with Thermo Scientific™ Accucore™ 150-C4 LC Columns. Based on Core Enhanced Technology, Thermo Scientific Accucore columns provide fast, high-resolution biomolecule separations without the elevated backpressures required by sub-2μm particles.

  • 150Å pore size solid core particles for fast biomoleucule separations
  • Superb resolution at low backpressures
  • Exceptionally rugged columns
  • Significantly lower hydrophobic retention than C18
  • Ideal for optimal retention of proteins and larger peptides

Ordering Information:

Other custom column dimensions are available. Please call your local Customer Service for more information.

Specifications

Specifications

Analytical Column
HPLC
14500 psi (1000 bar)
2 to 9
80 m²/g
L26
Accucore
2%
Yes
2.1 mm
30 mm
2.6 μm
150 Å
60°C
C4
Spherical, Solid Core, Ultrapure Silica
Reversed Phase
Reversed Phase
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.