Learn More
Thermo Scientific™ Accessories for the LP Vortex Mixer
Optimize flexibility and performance by using Thermo Scientific™ Accessories for LP Vortex Mixers.
Brand:  Thermo Scientific™ 88880120
Additional Details : Weight : 0.08730kg
 View more versions of this product
Product Code. 15268874
  • 15.80€ / Each
Estimated Shipment: 30-09-2024
Description
Optimize flexibility and performance by using Thermo Scientific™ Accessories for LP Vortex Mixers.
  •  Warranty: 2 years
Specifications
Specifications
Tube Holder
10 mm
Tube Holder with Insert Retainer (dia. 10mm)
Vortex Mixer
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.