Learn More
Digi-Sense™ Accessories for the 20250-40 Digi-Sense™ Temperature Humidity Touch-screen Recorder
The touch-screen reader has 8GB on board memory to provide virtually unlimited data logging capacity
Brand: Digi-Sense™ 20250-45
Additional Details : Weight : 0.01000kg
Description
- Includes A2LA accredited NIST-Traceable Calibration Certificate
- Measures, logs, and displays temperature, humidity, and dew point
- Removable sensor for local or remote measurements using extension cable included
- Wall mount or desk mount operation
- Resident self-booting software, no installation necessary
- Comes with NIST calibration
- Stored data downloads to computer via USB cable provided in CSV format
Services
2 Years
Specifications
Replacement Mounting Bracket | |
Temperature/RH Touch Screen Recorder with NIST Traceable Calibration (20250-40) |