Learn More
Cytiva Accessories for SE 250/260 Mini-vertical Gel Units: Alumina Notched Plate
Brand:  Cytiva SE202N-10
395.25 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 0.19200kg
 View more versions of this product
Product Code. 11735393
  • 422.00€ / Pack of 10
Estimated Shipment: 31-05-2024
Specifications
SE 250 Mini Vertical Gel Electrophoresis Unit
Alumina
8 cm
Notched Alumina Plate
10 cm