Learn More
Axygen™ Accessories for Horizontal Gel Box, 10-20 cm
For use with horizontal gel box, 10cm.
Brand:  Axygen HGB20-40MC-15
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 15543157
  • 74.20€ / 1 unit
Estimated Shipment: 19-07-2024
Description
Combs for 10-20 cm Axygen™ horizontal gel box
Specifications
Specifications
Horizontal Electrophoresis System Accessories
40-well Multi-Channel Compatible Comb, 1.5 mm thickness
1/Cs.
1.5 mm
Axygen Horizontal Gel Box, 20 cm
40
1/Pk.