Learn More
Axygen™ Accessories for Horizontal Gel Box, 10-20 cm
For use with horizontal gel box, 10cm.
Brand:  Axygen HGB20-30-1
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 15523157
  • 74.20€ / 1 unit
Estimated Shipment: 22-07-2024
Description
Combs for 10-20 cm Axygen™ horizontal gel box
Specifications
Specifications
Horizontal Electrophoresis System Accessories
30-well Comb, 1.0 mm thickness
1 mm
Axygen Horizontal Gel Box, 20 cm
30