Learn More
Merck Accessories for 47 mm All-glass Filter Holders Kits and MilliSolve Kit
Designed to replace parts contained in glass vacuum filtration kits or provide for an alternative configuration
Brand: Merck XX5014704
220.34 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.64080kg
Description
Designed to replace parts contained in glass vacuum filtration kits or provide for an alternative configuration
Specifications
Clear | |
Borosilicate Glass | |
500 mL |
Ground glass seal | |
Ground glass seal | |
47 mm |