Learn More
BINDER™ Access Port
For Use With: MKT 115
Brand:  BINDER™ 8012-1852
Additional Details : Weight : 0.00200kg
Product Code. 15963022
  • 2953.00€ / Each
Estimated Shipment: 01-08-2024
Description
  • Notch-type
  • For use in door
  • For use with Dynamic Climate Chambers
Specifications
Specifications
Notch-type access port - notch-type access port in door, 100 x 35 mm
BINDER Chambers MKT 115
Access Port
100 x 35 mm