Learn More
Fisherbrand™ ABS Plastic Blade
Product Code. 11928014 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Parafilm Sealing Film Dispenser
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11928014 Parafilm Sealing Film Dispenser Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11928014
  • ONLINE EXCLUSIVE PRICE
    6.70€
    5.78€ / Pack of 5
    Save 0.92€
    13% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
Replaces worn safety blade. Fisherbrand™ ABS Plastic Blade cuts sealing film without danger to fingers once properly installed in a sealing film dispenser.
For use with Parafilm™ Sealing Film Dispenser
Specifications
Product Type Replacement Blades
Color Clear
Material ABS
For Use With (Equipment) Parafilm Sealing Film Dispenser