Learn More
Esculape Laboratoire™ ABS First AID Kits
Brand: Esculape Laboratoire™ 720 01 01
Additional Details : Weight : 0.80000kg
Specifications
First Aid Kit | |
19 cm | |
ABS | |
1 Ea. |
17 cm | |
Blue | |
Gas band 3m x 7cm, Extensible web 3m x 7cm, 2 Alcoholic compresses, 2 Anti-compress shots, 2 Mosquito compress, 5 Sterile compresses 20 x 20, Guide first aid, 4 number of clips, Scissors, Pair of Vinyl gloves, Sterile pressure dressing 2.5m x 10cm, 2 ster | |
7 cm |