Learn More
Abnova™ CEN17q (FITC) FISH Probe
Labeled FISH probes as chromosomal markers using Fluorescent In Situ Hybridization Technique.
Brand: Abnova™ FC0136.20uL
Additional Details : Weight : 0.02000kg
Includes: FISH Hybridization Buffer (80μL)
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifications
Store at 4°C in the dark. | |
Fluorescent In Situ Hybridization (Cell) | |
17q11.2 | |
CEN17q (FITC) FISH Probe |
FITC | |
FISH Hybridization Buffer (80 uL) | |
CEN17q | |
20 μL |