Learn More
Abnova™ 19q Subtelomere (R6G) FISH Probe
Labeled FISH probes for identification of subtelomere aberrations using Fluorescent In Situ Hybridization Technique.
Brand: Abnova™ FE0173.20uL
Additional Details : Weight : 0.02000kg
Includes: FISH Hybridization Buffer (80μL)
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifications
Store at 4°C in the dark. | |
Fluorescent In Situ Hybridization (Cell) | |
19q Subtelomere | |
20 μL |
R6G | |
FISH Hybridization Buffer (80 uL) | |
19q Subtelomere(R6G) FISH Probe |