Learn More
AAV-MAX Transfection Kit, prototype
The Gibco AAV-MAX Transfection Kit, prototype, is an essential component of the AAV-MAX Helper-Free AAV production system.
Brand: Invitrogen A50518
Additional Details : Weight : 0.15000kg
Description
It is formulated for the transient transfection of Viral Production Cells 2.0. The kit includes a cationic lipid-based transfection reagent, as well as a transfection booster and production enhancer, designed to work together to produce adeno-associated virus (AAV) particles from Viral Production Cells 2.0 when cultured in Viral Production Medium.
Specifications
HEK293 | |
• 6 mL AAV-MAX Trasfection Reagent • 3 mL AAV-MAX Transfection Booster • 10 mL AAV-MAX Enhancer Store at 2–8°C. Do not freeze. |
|
Plasmid DNA | |
AAV Production | |
Wet Ice |
animal origin free, chemically defined | |
suspension | |
Lipid-based Transfection | |
AAV-MAX |
For Research Use Only. Not for use in diagnostic procedures. This product is a prototype and performance characteristics of this product have not been established. This product is currently undergoing real-time stability and shelf-life studies.