missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Solid Bottomed Microplates
For high-throughput screening and assay
Brand: Greiner Bio-One 651161
Additional Details : Weight : 0.80000kg
Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Ideally suited for agglutination tests
- No sharp corners, which means that they can be pipetted easily and cleanly
- Suitable for +/- analyses
- Solid bot
Specifications
No | |
Without Lid | |
Non Binding | |
Polystyrene | |
Sterile | |
234 μL | |
4800 x g (swinging bucket rotor) | |
100/Cs. | |
Chimney |
Standard | |
8 x 12 | |
Clear | |
96 | |
Non-treated | |
Conical Bottom (V-well) | |
100 | |
Clear |