Learn More
Greiner Bio-One 96-well CrystalQuick™ Crystallisation Microplates
For automatised applications. Greiner Bio-One 96-Well CrystalQuick™ Crystallisation Microplates are optimised for sealing with VIEWseal adhesive film.
Brand:  Greiner Bio-One 609871
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10145553
  • 1910.00€ / Pack of 80
Estimated Shipment: 02-07-2024
Description
  • Non-sterile
  • Polystyrene
  • Good optical properties
  • With crystallisation well per reservoir
  • Alphanumeric well coding
  • With CrystalDrop™ lid, experiments are possible with sitting drop and hanging drop metho
Specifications
Specifications
Standard
Inner bags of 20
LBR
3.9 μL
Non-sterile
Sitting drop applications with different well profiles and material properties
80/Cs.
8 x 12
Clear
Polystyrene
96
Square, Flat Bottom (F-well)
80
8 mm (Low profile)